3,5-Dimethylphenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid

129740-1G 129740-25G 129740-5G
BT129740-1GEA 46 USD
BT129740-1G BT129740-25G BT129740-5G
3,5-Dimethylphenylboronic acid ≥98%
3,5-Dimethylphenylboronic acid

CAS: 172975-69-8; MDL No: MFCD00185689 Solid; Molecular Formula: C8H11BO2 ; MW: 149.98 Melting Point: 261-265°


  • Solid

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 261…265 °C
MDL Number: MFCD00185689
CAS Number: 172975-69-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR