4-Fluoro-DL-phenylglycine ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Fluoro-DL-α-phenylglycine, 4-Fluorophenylglycine, H-DL-Phg(4-F)-OH

129695-1G 129695-5G
BT129695-1GEA 35 USD
BT129695-1G BT129695-5G
4-Fluoro-DL-phenylglycine ≥97%
4-Fluoro-DL-phenylglycine

CAS: 7292-73-1; EC No: 230-724-4; MDL No: MFCD00066444 Solid; Molecular Formula: C8H8FNO2; MW: 169.16


  • Solid

Formula: C₈H₈FNO₂
MW: 169.155 g/mol
MDL Number: MFCD00066444
CAS Number: 7292-73-1
EINECS: 230-724-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR