Fast blue BB salt

Supplier: BeanTown Chemical

Synonyms: 4-Benzoylamino-2,5-diethoxybenzenediazonium chloride hemi(zinc chloride) salt, 4-Amino-2,5-diethoxybenzanilide diazotated zinc double salt, Fast Blue BB Salt hemi(zinc chloride) salt

129675-1G 129675-5G
BT129675-1GEA 22 USD
BT129675-1G BT129675-5G
Fast blue BB salt
Fast blue BB salt

CAS: 5486-84-0; EC No: 226-817-4; MDL No: MFCD00074765 Powder; Molecular Formula: C34H36Cl4N6O6Zn; MW: 831.89 Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₃₄H₃₆Cl₄N₆O₆Zn
MDL Number: MFCD00074765
CAS Number: 5486-84-0
EINECS: 226-817-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR