Diammonium hydrogen orthophosphate ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

129665-1KG 129665-250G
BT129665-1KGEA 112.34 USD
BT129665-1KG BT129665-250G
Diammonium hydrogen orthophosphate ≥98.0% ACS
Diammonium hydrogen orthophosphate

CAS: 7783-28-0; EC No: 231-987-8; MDL No: MFCD00010891 Crystalline; Linear Formula: (NH4)2HPO4 ; MW: 132.06 Melting Point: 155° (decomposes)


  • Crystalline

Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185 (dec.)
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
Merck Index: 13,00546

Order Now

Specification Test Results

Assay 98.0% min
pH of 5% solution at 25°C 7.7-8.1
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.001%
Nitrate (NO3) ≤0.003%
Sulfate (SO4) ≤0.01%
Heavy metals (as Pb) ≤0.001%
Iron (Fe) ≤0.001%
Calcium (Ca) ≤0.001%
Magnesium (Mg) ≤0.0005%
Potassium (K) ≤0.005%
Sodium (Na) ≤0.005%
Ammonium hydroxide PPT ≤0.005%
Solubility Clear and colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR