1-Boc-indole ≥97%

Supplier: BeanTown Chemical

Synonyms: tert-Butyl 1-indolecarboxylate

129645-1G 129645-5G
BT129645-1GEA 48 USD
BT129645-1G BT129645-5G
1-Boc-indole ≥97%
1-Boc-indole

CAS: 75400-67-8; MDL No: MFCD02093939 Liquid; Molecular Formula: C13H15NO2; MW: 217.26 Boiling Point: 201°; Flash point: 71°C (160°F) Density (g/mL): 1.07; Refractive Index: 1.543


  • Refractive index: 1.543
  • Liquid

Formula: C₁₃H₁₅NO₂
MW: 217.27 g/mol
Boiling Pt: 201 °C
Density: 1.07 g/mL
Flash Pt: 71 °C (160 °F)
MDL Number: MFCD02093939
CAS Number: 75400-67-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR