5-Bromo-2-chlorobenzoic acid ≥98%
Supplier: BeanTown Chemical
129615-10G
129615-250G
129615-50G
BT129615-10GEA
44
USD
BT129615-10G
BT129615-250G
BT129615-50G
5-Bromo-2-chlorobenzoic acid ≥98%
5-Bromo-2-chlorobenzoic acid
CAS: 21739-92-4; EC No: 244-558-5; MDL No: MFCD00002415
Powder; Molecular Formula: C7H4BrClO2; MW: 235.46
Melting Point: 154-156°
- Powder
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol Melting Pt: 154…156 °C |
MDL Number:
MFCD00002415 CAS Number: 21739-92-4 EINECS: 244-558-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...