Brilliant green
Supplier: BeanTown Chemical
Synonyms:
Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
129590-100G
129590-25G
BT129590-100GEA
75
USD
BT129590-100G
BT129590-25G
Brilliant green
Brilliant green
CAS: 633-03-4; EC No: 211-190-1; MDL No: MFCD00011880; RTECS: BP6825000
Crystalline; Molecular Formula: C27H34N2O4S; MW: 485.63
Melting Point: 210° (decomposes)
- RTECS: BP6825000
- Crystalline
Formula:
C₂₇H₃₄N₂O₄S MW: 482.65 g/mol Melting Pt: 210 °C (dec.) |
MDL Number:
MFCD00011880 CAS Number: 633-03-4 EINECS: 211-190-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...