Acid red 27 ≥85-95% (dye content)

Supplier: BeanTown Chemical

Synonyms: Amaranth, Sodium 3-hydroxy-4-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulphonate, Bordeaux S, Food Red No. 2, Azo Rubine S, Wool Red 40 F, Naphthol Red

129580-100G 129580-500G 129580-50G
BT129580-100GEA 98.18 USD
BT129580-100G BT129580-500G BT129580-50G
Acid red 27 ≥85-95% (dye content)
Acid red 27

CAS: 915-67-3; EC No: 213-022-2; MDL No: MFCD00004076; RTECS: QJ6550000 Solid; Molecular Formula: C20H11N2Na3O10S3; MW: 604.47 Density (g/mL): 1.5


  • RTECS: QJ6550000
  • Solid

Formula: C₂₀H₁₄N₂O₁₀S₃
MW: 604.46 g/mol
Density: 1.5 g/mL
MDL Number: MFCD00004076
CAS Number: 915-67-3
EINECS: 213-022-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR