p-Xylene-2-sulfonic acid hydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
2,5-Dimethylbenzenesulphonic acid
129570-100G
129570-25G
129570-500G
BT129570-100GEA
142
USD
BT129570-100G
BT129570-25G
BT129570-500G
p-Xylene-2-sulfonic acid hydrate ≥99%
p-Xylene-2-sulfonic acid hydrate
CAS: 609-54-1; EC No: 210-193-5; MDL No: MFCD00151010
Solid; Molecular Formula: C8H10O3S·xH2O; MW: 186.23 (anhydrous)
Melting Point: 86°
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₈H₁₀O₃S MW: 186.23 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD00064858 CAS Number: 609-54-1 EINECS: 210-193-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...