3-(Hydroxymethyl)indole ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
129565-1G
129565-5G
BT129565-1GEA
23
USD
BT129565-1G
BT129565-5G
3-(Hydroxymethyl)indole ≥97%
3-(Hydroxymethyl)indole
CAS: 700-06-1; EC No: 211-836-2; MDL No: MFCD00005632; RTECS: NL9483000
Powder; Molecular Formula: C9H9NO; MW: 147.17
Melting Point: 96-99°
- RTECS: NL9483000
- Powder
Formula:
C₉H₉NO MW: 147.18 g/mol Melting Pt: 96…99 °C |
MDL Number:
MFCD00005632 CAS Number: 700-06-1 EINECS: 211-836-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...