Calcium hydrogen orthophosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic

129550-1KG 129550-250G 129550-5KG
BT129550-1KGEA 88.48 USD
BT129550-1KG BT129550-250G BT129550-5KG
Calcium hydrogen orthophosphate ≥98%
Calcium hydrogen orthophosphate

CAS: 7757-93-9; EC No: 231-826-1; MDL No: MFCD00010909 Powder ; Linear Formula: CaHPO4; MW: 136.06


  • Powder

Formula: CaHPO₄
MW: 136.06 g/mol
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR