Bis(4-nitrophenyl)carbonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Bis(4-nitrophenyl) carbonate

129545-25G 129545-5G
BT129545-25GEA 173 USD
BT129545-25G BT129545-5G
Bis(4-nitrophenyl)carbonate ≥98%
Bis(4-nitrophenyl)carbonate

CAS: 5070-13-3; EC No: 225-775-4; MDL No: MFCD00007322 Powder; Molecular Formula: C13H8N2O7; MW: 304.21 Melting Point: 136-139° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₁₃H₈N₂O₇
MW: 304.22 g/mol
Melting Pt: 136…139 °C
MDL Number: MFCD00007322
CAS Number: 5070-13-3
EINECS: 225-775-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR