Styrene-4-sulfonic acid sodium salt
Supplier: BeanTown Chemical
Synonyms:
Sodium 4-vinylbenzenesulphonate, 4-Vinylbenzenesulphonic acid sodium salt, Sodium-p-styrenesulphonate, p-Styrenesulphonic acid sodium salt
129510-250G
129510-50G
BT129510-250GEA
132
USD
BT129510-250G
BT129510-50G
Styrene-4-sulfonic acid sodium salt
Styrene-4-sulfonic acid sodium salt
CAS: 2695-37-6; EC No: 220-266-3; MDL No: MFCD00013379
Powder; Linear Formula: H2C=CHC6H4SO3Na; Molecular Formula: C8H7NaO3S; MW: 206.19
Melting Point:
- Air Sensitive, Light Sensitive
- Powder
Formula:
C₈H₇NaO₃S MW: 206.19 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00013379 CAS Number: 2695-37-6 EINECS: 220-266-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...