Benzoic anhydride ≥98%

Supplier: BeanTown Chemical

129490-1KG 129490-250G 129490-50G
BT129490-1KGEA 232 USD
BT129490-1KG BT129490-250G BT129490-50G
Benzoic anhydride ≥98%
Benzoic anhydride

CAS: 93-97-0; EC No: 202-291-1; MDL No: MFCD00003073 Crystalline; Molecular Formula: C14H10O3; MW: 226.23 Melting Point: 38-42°; Flash point: 110°C (230°F) Density (g/mL): 1.199 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Melting Pt: 38…42 °C
Density: 1.199 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00003073
CAS Number: 93-97-0
EINECS: 202-291-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR