(2-Chloroethoxy)benzene ≥97%

Supplier: BeanTown Chemical

Synonyms: ω-Chlorophenetole, 2-Chloroethyl phenyl ether, β-Chlorophenetole, 2-Phenoxyethyl chloride

129455-25G 129455-5G
BT129455-25GEA 134 USD
BT129455-25G BT129455-5G
(2-Chloroethoxy)benzene ≥97%
(2-Chloroethoxy)benzene

CAS: 622-86-6; EC No: 210-757-0; MDL No: MFCD00018946 Liquid; Molecular Formula: C88H9ClO; MW: 156.61 Boiling Point: 97-98°/15 mmHg; Flash point: 101°C (214°F) Density (g/mL): 1.129; Refractive Index: 1.534


  • Refractive index: 1.534
  • Liquid

Formula: C₆H₅OCH₂CH₂Cl
MW: 156.61 g/mol
Boiling Pt: 97…98 °C/15 mmHg
Density: 1.129 g/mL
Flash Pt: 101 °C (214 °F)
MDL Number: MFCD00018946
CAS Number: 622-86-6
EINECS: 210-757-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR