2-Chloronicotinamide ≥98%

Supplier: BeanTown Chemical

129445-1G 129445-25G 129445-5G
BT129445-1GEA 30 USD
BT129445-1G BT129445-25G BT129445-5G
2-Chloronicotinamide ≥98%
2-Chloronicotinamide

CAS: 10366-35-5; EC No: 233-808-9; MDL No: MFCD00006237 Crystalline; Molecular Formula: C6H5ClN2O; MW: 156.57 Melting Point: 164-167°


  • Crystalline

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Melting Pt: 164…167 °C
MDL Number: MFCD00006237
CAS Number: 10366-35-5
EINECS: 233-808-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR