Omeprazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
129415-1G
129415-5G
BT129415-1GEA
49.05
USD
BT129415-1G
BT129415-5G
Omeprazole ≥98%
Omeprazole
CAS: 73590-58-6; MDL No: MFCD00083192; RTECS: DD9087000
Crystalline; Molecular Formula: C17H19N3O3S; MW: 345.42
Melting Point: 156°
- RTECS: DD9087000
- Crystalline
Formula:
C₁₇H₁₉N₃O₃S MW: 345.42 g/mol Melting Pt: 156 °C |
MDL Number:
MFCD00083192 CAS Number: 73590-58-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...