4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99%
Supplier: BeanTown Chemical
Synonyms:
5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane
129410-100G
129410-25G
129410-5G
BT129410-100GEA
533.34
USD
BT129410-100G
BT129410-25G
BT129410-5G
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
CAS: 1107-00-2; EC No: 214-170-0; MDL No: MFCD00039143
Crystalline; Molecular Formula: C19H6F6O6; MW: 444.24
Melting Point: 244-247°
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₈H₄O₃ MW: 444.25 g/mol Melting Pt: 244…247 °C |
MDL Number:
MFCD00039143 CAS Number: 1107-00-2 EINECS: 214-170-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...