Iron(III) meso-tetraphenylporphine chloride

Supplier: BeanTown Chemical

Synonyms: Iron(III) Tetraphenylporphyrin chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride, meso-Tetraphenylporphyrin iron(III) chloride, iron(III) chloride TPP, 21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride

129390-1G 129390-250MG
BT129390-1GEA 221 USD
BT129390-1G BT129390-250MG
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride

CAS: 16456-81-8; MDL No: MFCD00012152 Crystalline; Molecular Formula: C44H28ClFeN4; MW: 704.04


  • Crystalline

Formula: C₄₄H₂₈ClFeN₄
MW: 704.02 g/mol
MDL Number: MFCD00012152
CAS Number: 16456-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR