L(+)-Isoleucine ≥99%
Supplier: BeanTown Chemical
129375-10G
129375-50G
BT129375-10GEA
39.23
USD
BT129375-10G
BT129375-50G
L(+)-Isoleucine ≥99%
L(+)-Isoleucine
Formula:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Melting Pt: 288 °C (dec.) |
MDL Number:
MFCD00064222 CAS Number: 73-32-5 EINECS: 200-798-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...