Antimony(III) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

129365-250G 129365-50G
BT129365-250GEA 560 USD
BT129365-250G BT129365-50G
Antimony(III) oxide ≥99.9%
Antimony(III) oxide

CAS: 1309-64-4; EC No: 215-175-0; MDL No: MFCD00011214; RTECS: CC5650000 25mm & Down Sintered Lump ; Linear Formula: Sb2O3; MW: 291.52 Melting Point: 655°; Boiling Point: 1550° (sublimes)


  • RTECS: CC5650000
  • 25mm & Down Sintered Lump

Formula: O₃Sb₂
MW: 291.52 g/mol
Boiling Pt: 1425 (subl.)
Melting Pt: 654
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1 III
Merck Index: 13,00717

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR