Dysprosium ≥99.5% (trace metals basis excluding Ta), rods Ø 6.35 mm

Supplier: BeanTown Chemical

129345-10MM 129345-25MM 129345-50MM
BT129345-10MMEA 147 USD
BT129345-10MM BT129345-25MM BT129345-50MM
Dysprosium ≥99.5% (trace metals basis excluding Ta), rods Ø 6.35 mm
Dysprosium

CAS: 7429-91-6; EC No: 231-073-9; MDL No: MFCD00010982 Solid, cast rod, diameter is approximate; Molecular Formula: Dy; MW: 162.50 Melting Point: 1412°; Boiling Point: 2567° Density (g/mL): 8.559


  • Solid, cast rod, diameter is approximate

Formula: Dy
MW: 162.5
Boiling Pt: 2567 °C
Melting Pt: 1412 °C
Density: 8.559 g/mL
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR