Diethyl propylphosphonate ≥97%

Supplier: BeanTown Chemical

Synonyms: Diethyl 1-propylphosphonate

129340-10G 129340-250G 129340-50G
BT129340-10GEA 49 USD
BT129340-10G BT129340-250G BT129340-50G
Diethyl propylphosphonate ≥97%
Diethyl propylphosphonate

CAS: 18812-51-6; MDL No: MFCD00015139 Liquid; Molecular Formula: C7H17O3P; MW: 180.19 Boiling Point: 94-96°/12 mmHg


  • Liquid

Formula: C₇H₁₇O₃P
MW: 180.19 g/mol
Boiling Pt: 94…96 °C/12 mmHg
MDL Number: MFCD00015139
CAS Number: 18812-51-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR