Linoleic acid ≥95%

Supplier: BeanTown Chemical

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

129310-100G 129310-25G 129310-5G
BT129310-100GEA 437 USD
BT129310-100G BT129310-25G BT129310-5G
Linoleic acid ≥95%
Linoleic acid

CAS: 60-33-3; EC No: 200-470-9; MDL No: MFCD00064241; RTECS: RF9990000 Liquid; Molecular Formula: C18H32O2; MW: 280.45 Melting Point: -5°; Boiling Point: 229-230°/16 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.902; Refractive Index: 1.466 Air Sensitive


  • RTECS: RF9990000
  • Refractive index: 1.466
  • Air Sensitive
  • Liquid

Formula: C₁₈H₃₂O₂
Boiling Pt: 229…230 °C/16 mmHg
Melting Pt: –5 °C
Density: 0.902 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR