Titanium(IV) oxide rutile ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Pigment White 6, Rutile, Titanium dioxide rutile

129300-10G 129300-250G 129300-50G
BT129300-10GEA 81 USD
BT129300-10G BT129300-250G BT129300-50G
Titanium(IV) oxide rutile ≥99.99% (trace metals basis)
Titanium(IV) oxide rutile

CAS: 1317-80-2; EC No: 215-282-2; MDL No: MFCD00011269; RTECS: XR2275000 Powder; Linear Formula: TiO2; MW: 79.90 Melting Point: 1830-1850°; Boiling Point: 2500-3000° Density (g/mL): 4.23


  • RTECS: XR2275000
  • Powder

Formula: O₂Ti
MW: 79.88 g/mol
Boiling Pt: 2500…3000 °C
Melting Pt: 1830…1850 °C
Density: 4.23 g/mL
MDL Number: MFCD00011269
CAS Number: 1317-80-2
EINECS: 215-282-2

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR