2-Bromomesitylene ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene

129265-250G 129265-50G
BT129265-250GEA 180 USD
BT129265-250G BT129265-50G
2-Bromomesitylene ≥99%
2-Bromomesitylene

CAS: 576-83-0; EC No: 209-405-9; MDL No: MFCD00000073 Liquid; Molecular Formula: C9H11Br; MW: 199.09 Melting Point: 2°; Boiling Point: 225°; Flash point: 96°C (205°F) Density (g/mL): 1.301; Refractive Index: 1.552 Light Sensitive


  • Refractive index: 1.552
  • Light Sensitive
  • Liquid

Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 225 °C
Melting Pt: 2 °C
Density: 1.301 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00000073
CAS Number: 576-83-0
EINECS: 209-405-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR