Magnesium thiosulphate hexahydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Magnesium hyposulphite hexahydrate
129235-100G
129235-500G
BT129235-100GEA
40
USD
BT129235-100G
BT129235-500G
Magnesium thiosulphate hexahydrate ≥99%
Magnesium thiosulphate hexahydrate
CAS: 13446-30-5; EC No: 233-340-5; MDL No: MFCD00169871; RTECS: OM4610000
Crystalline; Molecular Formula: MgO3S2·6H2O ; MW: 244.53
Density (g/mL): 1.818
- RTECS: OM4610000
- Crystalline
Formula:
MgO₃S₂·6H₂O Density: 1.818 g/mL |
MDL Number:
MFCD00169871 CAS Number: 13446-30-5 EINECS: 233-340-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...