Silicone oil for oil baths
Supplier: BeanTown Chemical
Synonyms:
Polyphenyl-methylsiloxane
129230-100G
129230-2.5KG
129230-500G
BT129230-100GEA
47.77
USD
BT129230-100G
BT129230-2.5KG
BT129230-500G
Silicone oil for oil baths
Silicone oil
CAS: 63148-62-9; MDL No: MFCD00132673; RTECS: JT6484500
Oil; Linear Formula: [-Si(CH3)2O-]n
Melting Point: -55°; Flash point: 316°C (601°F)
Density (g/mL): 0.967; Refractive Index: 1.403
- RTECS: JT6484500
- Refractive index: 1.403
- Oil
Melting Pt:
–55 °C Density: 0.967 g/mL Flash Pt: 316 °C (601 °F) |
MDL Number:
MFCD00132673 CAS Number: 68083-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...