4,6-Dichloro-5-nitropyrimidine ≥98%
Supplier: BeanTown Chemical
129205-1G
129205-25G
129205-5G
BT129205-1GEA
69
USD
BT129205-1G
BT129205-25G
BT129205-5G
4,6-Dichloro-5-nitropyrimidine ≥98%
4,6-Dichloro-5-nitropyrimidine
CAS: 4316-93-2; EC No: 224-340-6; MDL No: MFCD00006107; RTECS: UV8258000
Crystalline; Molecular Formula: C4HCl2N3O2; MW: 193.98
Melting Point: 100-103°
- RTECS: UV8258000
- Crystalline
Formula:
C₄HCl₂N₃O₂ Melting Pt: 100…103 °C |
MDL Number:
MFCD00006107 CAS Number: 4316-93-2 EINECS: 224-340-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...