Rubidium chloride ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Rubidiumchloride

129190-10G 129190-50G
BT129190-10GEA 125.68 USD
BT129190-10G BT129190-50G
Rubidium chloride ≥99.5% (trace metals basis)
Rubidium chloride

CAS: 7791-11-9; EC No: 232-240-9; MDL No: MFCD00011187; RTECS: VL8575000 -20 Mesh Powder; Linear Formula: RbCl; MW: 120.92 Melting Point: 715°; Boiling Point: 1390° Density (g/mL): 2.8; Refractive Index: 1.493 Hygroscopic


  • RTECS: VL8575000
  • Refractive index: 1.493
  • Hygroscopic
  • -20 Mesh Powder

Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390 °C
Melting Pt: 715 °C
Density: 2.8 g/mL
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR