3-Hydroxy-2-nitropyridine

Supplier: BeanTown Chemical

Synonyms: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol

129175-10G 129175-250G 129175-50G
BT129175-10GEA 54 USD
BT129175-10G BT129175-250G BT129175-50G
3-Hydroxy-2-nitropyridine
3-Hydroxy-2-nitropyridine

CAS: 15128-82-2; EC No: 239-191-2; MDL No: MFCD00006258; RTECS: UU7715000 Solid; Molecular Formula: C5H4N2O3 ; MW: 140.10 Melting Point: 69-71°


  • RTECS: UU7715000
  • Solid

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 70
MDL Number: MFCD00006258
CAS Number: 15128-82-2
EINECS: 239-191-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR