2,3,5-Trichloroisonicotinic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,3,5-Trichloropyridine-4-carboxylic acid

129150-1G 129150-250MG 129150-5G
BT129150-1GEA 71 USD
BT129150-1G BT129150-250MG BT129150-5G
2,3,5-Trichloroisonicotinic acid ≥98%
2,3,5-Trichloroisonicotinic acid

CAS: 406676-18-4 Solid; Molecular Formula: C6H2Cl3NO2; MW: 226.44


  • Solid

Formula: C₆H₂Cl₃NO₂
MW: 226.44 g/mol
MDL Number: MFCD11044390
CAS Number: 406676-18-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR