Nickel ≥99.5% (trace metals basis), plate, annealed, Thickness 6.35 mm
Supplier: BeanTown Chemical
129145-100X100
129145-50X50MM
BT129145-100X1EA
439.96
USD
BT129145-100X1
BT129145-50X50
Nickel ≥99.5% (trace metals basis), plate, annealed, Thickness 6.35 mm
Nickel
CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137
Solid; Molecular Formula: Ni; MW: 58.69
Melting Point: 1453°; Boiling Point: 2732°
Density (g/mL): 8.9
- Solid
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.9 g/mL |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...