Molybdenum ≥99.95% (trace metals basis), rods Ø 6.4 mm

Supplier: BeanTown Chemical

129130-250G 129130-50G
BT129130-250GEA 200 USD
BT129130-250G BT129130-50G
Molybdenum ≥99.95% (trace metals basis), rods Ø 6.4 mm
Molybdenum

CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465; RTECS: QA4680000 Solid; Molecular Formula: Mo; MW: 95.94 Melting Point: 2617°; Boiling Point: 4612° Density (g/mL): 10.3


  • RTECS: QA4680000
  • Solid

Formula: Mo
Boiling Pt: 4612 °C
Melting Pt: 2617 °C
Density: 10.3 g/mL
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR