Manganese(IV) oxide, Technical Grade, activated

Supplier: BeanTown Chemical

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

129120-100G 129120-500G
BT129120-100GEA 38.62 USD
BT129120-100G BT129120-500G
Manganese(IV) oxide, Technical Grade, activated
Manganese(IV) oxide

CAS: 1313-13-9; EC No: 215-202-6; MDL No: MFCD00003463; RTECS: OP0350000 Powder; Molecular Formula: MnO2; MW: 86.94 Melting Point: 535° (decomposes) Density (g/mL): 5.026


  • RTECS: OP0350000
  • Powder

Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C (dec.)
Density: 5.026 g/mL
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR