6-Methyl-2-thiouracil ≥98%

Supplier: BeanTown Chemical

Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin

129060-1KG 129060-250G 129060-50G
BT129060-1KGEA 174 USD
BT129060-1KG BT129060-250G BT129060-50G
6-Methyl-2-thiouracil ≥98%
6-Methyl-2-thiouracil

CAS: 56-04-2; EC No: 200-252-3; MDL No: MFCD00006040; RTECS: YR0875000 Powder; Molecular Formula: C5H6N2OS; MW: 142.18 Melting Point: 330° (decomposes)


  • RTECS: YR0875000
  • Powder

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 330 °C (dec.)
MDL Number: MFCD00006040
CAS Number: 56-04-2
EINECS: 200-252-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR