Diboron trioxide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Boron oxide, Boric anhydride
129035-10KG
129035-2KG
129035-500G
BT129035-10KGEA
307
USD
BT129035-10KG
BT129035-2KG
BT129035-500G
Diboron trioxide ≥98%
Diboron trioxide
CAS: 1303-86-2; EC No: 215-125-8; MDL No: MFCD00011315; RTECS: ED7900000
-60 Mesh Powder; Molecular Formula: B2O3; MW: 69.62
Melting Point: 45°
Density (g/mL): 2.46
Hygroscopic
- RTECS: ED7900000
- Hygroscopic
- -60 Mesh Powder
Formula:
B₂O₃ MW: 69.62 g/mol Melting Pt: 45 °C Density: 2.46 g/mL |
MDL Number:
MFCD00011315 CAS Number: 1303-86-2 EINECS: 215-125-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...