3,5-Dimethylbenzonitrile ≥98%

Supplier: BeanTown Chemical

129020-1G 129020-5G
BT129020-1GEA 31 USD
BT129020-1G BT129020-5G
3,5-Dimethylbenzonitrile ≥98%
3,5-Dimethylbenzonitrile

CAS: 22445-42-7; EC No: 245-001-9; MDL No: MFCD00060670 Solid; Linear Formula: NCC6H3(CH3)2; Molecular Formula: C9H9N; MW: 131.17 Boiling Point: 243°


  • Solid

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 243 °C
MDL Number: MFCD00060670
CAS Number: 22445-42-7
EINECS: 245-001-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR