DL-Alanine ≥99%

Supplier: BeanTown Chemical

Synonyms: (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine

129005-1KG 129005-250G 129005-5KG
BT129005-1KGEA 86 USD
BT129005-1KG BT129005-250G BT129005-5KG
DL-Alanine ≥99%
DL-Alanine

CAS: 302-72-7; EC No: 206-126-4; MDL No: MFCD00064408; RTECS: AY2980000 Powder; Linear Formula: CH3CH(NH2)COOH; Molecular Formula: C3H7NO2; MW: 89.09 Melting Point: 289° (decomposes)


  • RTECS: AY2980000
  • Powder

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79 (dec.)
MDL Number: MFCD00064408
CAS Number: 302-72-7
EINECS: 206-126-4
Merck Index: 13,00203

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR