(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

129000-1G 129000-5G
BT129000-1GEA 61 USD
BT129000-1G BT129000-5G
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid

CAS: 7326-19-4; EC No: 230-803-3; MDL No: MFCD00078062 Crystalline; Molecular Formula: C9H10O3 ; MW: 166.17 Melting Point: 122-124° Optical Rotation: [α]20/D +19°, c = 1 in ethanol


  • Optical rotation: [α]20/D +19°, c = 1 in ethanol
  • Crystalline

Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00078062
CAS Number: 7326-19-4
EINECS: 230-803-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR