L(+)-Leucine ≥99%

Supplier: BeanTown Chemical

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

128980-100G 128980-2.5KG 128980-500G
BT128980-100GEA 63.64 USD
BT128980-100G BT128980-2.5KG BT128980-500G
L(+)-Leucine ≥99%
L(+)-Leucine

CAS: 61-90-5; EC No: 200-522-0; MDL No: MFCD00002617; RTECS: OH2850000 Powder; Molecular Formula: C6H13NO2; MW: 131.17 Optical Rotation: [α]20/D +15.5±0.5°, c = 5% in 5M HCl


  • RTECS: OH2850000
  • Optical rotation: [α]20/D +15.5±0.5°, c = 5% in 5M HCl
  • Powder

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR