cis-N-Boc-4-cyano-L-proline methyl ester ≥97%

Supplier: BeanTown Chemical

Synonyms: Boc-cis-4-CN-Pro-OMe, (2S,4S)-1-tert-Butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, 1-tert-Butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate

128970-1G 128970-250MG
BT128970-1GEA 1028 USD
BT128970-1G BT128970-250MG
cis-N-Boc-4-cyano-L-proline methyl ester ≥97%
cis-N-Boc-4-cyano-L-proline methyl ester

CAS: 487048-28-2; MDL No: MFCD03787928 Solid; Molecular Formula: C12H18N2O4; MW: 254.29


  • Solid

Formula: C₁₂H₁₈N₂O₄
MDL Number: MFCD03787928
CAS Number: 487048-28-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR