γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99%

Supplier: BeanTown Chemical

Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

128910-100G 128910-25G 128910-500G
BT128910-100GEA 90.04 USD
BT128910-100G BT128910-25G BT128910-500G
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99%
γ-(N-Morpholino)propanesulphonic acid (MOPS)

CAS: 1132-61-2; EC No: 214-478-5; MDL No: MFCD00006183; RTECS: QE9104530 Powder; Molecular Formula: C7H15NO4S; MW: 209.27 Melting Point: 277-280°; Flash point: 110°C (230°F)


  • RTECS: QE9104530
  • Powder

Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: 277…280 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR