Calcium titanate ≥99%

Supplier: BeanTown Chemical

Synonyms: Calcium titanate (IV), Calcium titanate, Calcium titanium oxide

128890-2KG 128890-500G
BT128890-2KGEA 306 USD
BT128890-2KG BT128890-500G
Calcium titanate ≥99%
Calcium titanate

CAS: 12049-50-2; EC No: 234-988-1; MDL No: MFCD00015981 -325 Mesh Powder; Linear Formula: CaTiO3; MW: 135.94 Density (g/mL): 4.1


  • -325 Mesh Powder

Formula: CaO₃Ti
MW: 135.96 g/mol
Density: 4.1 g/mL
MDL Number: MFCD00015981
CAS Number: 12049-50-2
EINECS: 234-988-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR