Toluene-2-sulfonamide ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide
128865-10G
128865-50G
BT128865-10GEA
37
USD
BT128865-10G
BT128865-50G
Toluene-2-sulfonamide ≥99%
Toluene-2-sulphonamide
CAS: 88-19-7; EC No: 200-741-1; MDL No: MFCD00007934; RTECS: XT5075000
Powder; Linear Formula: CH3C6H4SO2NH2; Molecular Formula: C7H9NO2S; MW: 171.22
Melting Point: 156-158°; Flash point: 178°C (352°F)
- RTECS: XT5075000
- Powder
Formula:
C₇H₉NO₂S MW: 171.22 g/mol Melting Pt: 156…158 °C Flash Pt: 178 °C (352 °F) |
MDL Number:
MFCD00007934 CAS Number: 88-19-7 EINECS: 200-741-1 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...