3-Fluoro-4-morpholinoaniline ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-(2-Fluoro-4-aminophenyl)morpholine
128845-25G
128845-5G
BT128845-25GEA
216
USD
BT128845-25G
BT128845-5G
3-Fluoro-4-morpholinoaniline ≥98%
3-Fluoro-4-morpholinoaniline
CAS: 93246-53-8; MDL No: MFCD02571270
Solid; Molecular Formula: C10H13FN2O; MW: 196.22
Melting Point: 121-123°; Boiling Point: 365°
- Solid
Formula:
C10H13FN2O Boiling Pt: 365 °C Melting Pt: 121…123 °C |
MDL Number:
MFCD02571270 CAS Number: 93246-53-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...