Diyttrium trioxide ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical

Synonyms: Yttrium oxide, Yttrium (III) oxide

128815-1KG 128815-250G 128815-50G
BT128815-1KGEA 578 USD
BT128815-1KG BT128815-250G BT128815-50G
Diyttrium trioxide ≥99.9% (trace rare earth metals basis)
Diyttrium trioxide

CAS: 1314-36-9; EC No: 215-233-5; MDL No: MFCD00011473 Powder; Linear Formula: Y2O3; MW: 225.81 Melting Point: 2410°; Boiling Point: 4300° Density (g/mL): 5.01 Air Sensitive


  • Air Sensitive
  • Powder

Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300 °C
Melting Pt: 2410 °C
Density: 5.01 g/mL
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR