Tin(IV) isopropoxide isopropanol adduct ≥98%
Supplier: BeanTown Chemical
128775-1G
128775-5G
BT128775-1GEA
60
USD
BT128775-1G
BT128775-5G
Tin(IV) isopropoxide isopropanol adduct ≥98%
Tin(IV) isopropoxide isopropanol adduct
CAS: 132951-93-0; MDL No: MFCD00145408
Solid; Linear Formula: Sn[OCH(CH3)2]4·C3H7OH ; Molecular Formula: C12H28O4Sn·C3H8O ; MW: 415.16
Melting Point: 130° (decomposes); Boiling Point: 131°/1.6 mmHg
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
Sn[OCH(CH₃)₂]₄·C₃H₇OH MW: 414.15 g/mol Boiling Pt: 131 °C/1.6 mmHg Melting Pt: 130 °C (dec.) |
MDL Number:
MFCD00145408 CAS Number: 132951-93-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...