Iron(II) ethylenediammonium sulfate tetrahydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ferrous Ethylenediamine Sulphate
128745-100G
128745-25G
BT128745-100GEA
86
USD
BT128745-100G
BT128745-25G
Iron(II) ethylenediammonium sulfate tetrahydrate ≥99%
Iron(II) ethylenediammonium sulfate tetrahydrate
CAS: 34962-29-3; EC No: 264-357-6; MDL No: MFCD00082474
Crystalline; Molecular Formula: C2H10FeN2O8S2·4H2O ; MW: 382.14
Formula:
C₂H₈N₂ MW: 310.07 g/mol |
MDL Number:
MFCD00082474 CAS Number: 113193-60-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...