Didodecyldimethylammonium bromide ≥98%
Supplier: BeanTown Chemical
128730-1G
128730-5G
BT128730-1GEA
24.23
USD
BT128730-1G
BT128730-5G
Didodecyldimethylammonium bromide ≥98%
Didodecyldimethylammonium bromide
CAS: 3282-73-3; EC No: 221-923-7; MDL No: MFCD00041969
Powder; Linear Formula: [CH3(CH2)11]2N(CH3)2Br; Molecular Formula: C26H56BrN ; MW: 462.63
Melting Point: 157-162°
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₂₆H₅₆BrN Melting Pt: 157…162 °C |
MDL Number:
MFCD00041969 CAS Number: 3282-73-3 EINECS: 221-923-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...